Bellow connectors with GL threads, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H902-15 H902-10 H902-05 H902-03 H902-04
BOHLH902-15EA 1740 DKK
BOHLH902-15 BOHLH902-10 BOHLH902-05 BOHLH902-03 BOHLH902-04
Bellow connectors with GL threads, BOLA
Fittings og stik
PTFE/FPM and PS.
  • Temperature resistance: from −200 to +250 °C
  • Chemical resistance: universal
  • Vacuum: suitable
Two screw caps made up of PPS with GL thread and PTFE/FPM gasket. Bellow with sharp folds made of PTFE. Bellow can be bent up to an angle of 120°, flexible and strainless connection.