Funnels with GL thread or ground joint
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D738-12 D738-22 D738-42 D739-22 D738-52
BOHLD738-12EA 1490 DKK
BOHLD738-12 BOHLD738-22 BOHLD738-42 BOHLD739-22 BOHLD738-52
Funnels with GL thread or ground joint
Tragte Pulvertragte
Powder funnels, made of borosilicate glass with outlet pipe of PTFE, connection with GL screw caps of PPS or ground joint.
  • The glass funnel can be fixed in any position
  • Used for reaction vessels with GL threaded connections or grounding sleeves
  • Temperature resistance −20 to +200 °C
  • Universal chemical resistance