Vial racks, Chromacol™
Leverandør: CHROMACOL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B-100 T-15/302 T-15/308 T-180 T-28
548-1269EA 1220 DKK
548-1269 548-1322 549-0182 548-1324 548-1327
Vial racks, Chromacol™
Stativer Stativer til Kromatografirør
These vial racks are chemically resistant and fairly robust for a safe working position on the lab bench and during transport.
  • Ideal for space saving storage in fridges
  • Temperature resistant from −90 to 121 °C (except foam)