Drum thieves, Wheaton®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
885300 885302 885303
WHEA885300EA 4690 DKK
WHEA885300 WHEA885302 WHEA885303
Drum thieves, Wheaton®
Prøvetagere Væskeprøvetagere
Drum thieves made of soda lime glass for quick disposal.
  • Conventional open end drum thieves
  • Just insert empty tube in drum
  • Hold thumb over top and withdraw