Storage units, MagLab™ series
Leverandør: HEATHROW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HS23500 HS23502 HS23503 HS23504
211-0382EA 933 DKK
211-0382 211-0384 211-0385 211-0386
Storage units, MagLab™ series
Bokse Opbevaringsbokse
The MagLab™ series allows common laboratory products to be organised off the benchtop.
ABS, coloured.
  • MagRack™ holds four 50 ml tubes and nine 15 ml tubes
  • MagWipe™ holds a standard (11×21 cm) Kimwipes® box
  • MagPette™ holds two pipettors (fits most popular styles)
  • MagBox™ holds miscellaneous accessories
Available individually or as a set of four, each unit is secured by magnets that hold onto most metal surfaces.