Transport and Storage Insulation Boxes
Leverandør: KGW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12620 12614-130 12615-160 12623
KGWA12620EA 0 DKK
KGWA12620 KGWA12614-130 KGWA12615-160 KGWA12623
Transport and Storage Insulation Boxes
Bokse Kølere/Frysebokse/Poser
Large insulation COB boxes are made of monobloc polyethylene.
  • Low own weight
  • Temperature ranges from −80 to +80 °C
  • Polyurethane foam insulation core 60 to 90 mm thick
  • With lockable lid and handle for easy carrying
Ideal for storage and transport of temperature sensitive goods such as dry ice, biological samples and food.
Certificeringer: Meets Euro-standard IN-IR-RRC.