VWR®, Bottles, Wide Neck, HDPE, Rectangular or Square, with Screw Cap
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
215-1904EA 186 DKK
215-1904 215-1905 215-1906 215-1907 215-1908 215-1909
VWR®, Bottles, Wide Neck, HDPE, Rectangular or Square, with Screw Cap
Flasker Plastflasker
HDPE.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof
For collecting, storing and shipping samples, or storing liquids and dry substances. Space saving design with rectangular or square base. 2 and 4 litre bottles have integrated handle design.