VWR®, Crucibles, PTFE
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216-0450EA 195 DKK
216-0450 216-0451 216-0452 216-0453
VWR®, Crucibles, PTFE
Digler
Isostatically moulded in pure PTFE, FDA approved.
  • Chemically resistant and inert
  • Crack-free
  • Withstand maximum temperature of 280 °C