VWR® Thermotech™, Beakers, Low Form, Heatable
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
213-0272EA 671 DKK
213-0272 213-0273 213-0271
VWR® Thermotech™, Beakers, Low Form, Heatable
Bægre
PTFE, white with black PTFE carbon base.
  • Totally inert pure PTFE inner surface
  • Heatable to 270 °C without distortion
  • Improved heat transfer
Features the unique combination of a pure PTFE body with a specially formulated stabilised PTFE carbon base as a single moulding.