Tubing clamps, swing jaw, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H182210000 H182220000
129-2233EA 1550 DKK
129-2233 129-2234
Tubing clamps, swing jaw, SP Bel-Art
Klemmer Rørklemmer
Zinc plated steel. Swing jaw clamps for use with all flexible tubing.
  • Attach at any point on tubing without disconnecting tubing
  • Zinc plated steel is chemical and corrosion resistant
  • Knurled nut applies clamping force through smooth clamping surfaces
  • Autoclavable