Laboratory coats, PE laminate, 4440
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4440XL 4440S 4440M 4440L 44404XL 44403XL 44402XL
113-7866EA 4530 DKK
113-7866 113-6252 113-6251 113-6250 113-6249 113-6248 113-6247
Laboratory coats, PE laminate, 4440
Engangsbeklædning
These disposable coats made of microporous laminate provide protection against non hazardous dusts and non hazardous light liquid splashes.
  • Knit cuffs for enhanced fit and comfortable wear
  • Zip closure for convenience
  • Low-linting properties, anti-static treatment on both sides
  • Lightweight microporous material to help reduce heat build-up
Typical applications include general maintenance, electronics manufacturing, food and beverage manufacturing, catering, the beauty industry and laboratories.
Colour: White