Protective coveralls, 4565
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4565M 4565XXL 4565L 45654XL 45653XL 4565S 7100256062 7100256143
113-5425EA 226 DKK
113-5425 MMMA4565XXL 113-5424 MMMA45654XL MMMA45653XL MMMA4565S MMMA7100256062 MMMA7100256143
Protective coveralls, 4565
Engangsbeklædning
3M™ Protective Coverall 4565 has desirable comfort features and provides a strong barrier to certain chemicals, liquid sprays, and particles. These coveralls are non-microporous to offer chemical protection with permeation tested resistance, and they are certified to EN 14126 infective agents.
  • Type 4/5/6 coverall designed to offer a very good barrier to hazardous particles and limited liquid splash and spray
  • Made of bicomponent, non-woven with ultra-low linting PE laminate for a noticeably comfortable drape
  • 3-panel hood, 2-way zip, and storm flap with sealable tape
  • Elasticated back and ankles to reduce the risk of skin exposure
  • Taped seams across the suit for increased hold-out and strength, reducing the risk of tearing and exposure
  • Protection against EN 14126 Biological agents
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold-out to class 2
3M™ Protective Coverall 4565 provides a superior barrier to hazardous particles, limited liquid splashes, and sprays to Type 4. The fresh white 4565 coverall has recognisable red taped seams and meets a variety of standards, including chemical protective clothing including EN 14126 Infective agents, EN 1073-2 Nuclear particulate resistance, EN 1149, EN ISO 13688, Type 6 Liquid Splash EN 13034, and Type 5 Dust EN ISO 13982-1.
Taped seams reduce skin exposure points through tearing and reduced risk of inward leakage by chemicals or particles. The coverall also offers protection to class 2 in Particulate Radioactive Contamination EN1073-2, meaning it has very good particle hold-out as well as liquid spray resistance. Made with a non-microporous PE laminate, this garment offers chemical protection with permeation tested resistance against certain chemicals. Knitted cuffs, and elasticated waist and ankles make the wearer more comfortable and increase freedom of movement, while the ultra-low linting material reduces environmental contamination. The 3-panel hood design is compatible with additional PPE. The 2-way zips and fully sealable storm flaps reduce the risk of skin exposure and protect from contaminants. This coverall also has an Anti-static treatment meeting EN 1149, which reduces static discharge and reduces the risk of spark and ignition.
Certificeringer: CE Category III. EN 1073-2 Particulate Radioactive Contamination. EN 1149-5 - Electrostatic Properties. EN 13034:2005+A1:2009 Type 6 Liquid Splash. EN 14126 - Infective Agents (Biohazard). EN 14605:2005+A1:2009 Type 4 Liquid Spray. EN ISO 13982-1:2004 Type 5 Hazardous Dusts.