Chemical Resistant Overalls, 4545
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
45452XL 45453XL 45454XL 4545L 4545M 4545XL 4545S
MMMA45452XLEA 2900 DKK
MMMA45452XL MMMA45453XL MMMA45454XL MMMA4545L MMMA4545M MMMA4545XL MMMA4545S
Chemical Resistant Overalls, 4545
Engangsbeklædning
These disposable overalls are made of a laminated microporous material consisting of polyethylene and polyester. The garments provide an excellent barrier against hazardous dusts (type 5) and certain light liquid splashes (type 6). Certified to also protect against radioactive particles and biological agents and reduce build up of static charge.
  • Soft, well-draping material
  • Elasticated waist and ankles for freedom of movement and knitted cuffs for extra comfort
  • Three-panel hood design for a better fit and compatibility with other PPE
  • Two-way zip and sealable storm flap for enhanced protection
  • Low-linting properties
  • Reinforced gusset
Typical applications may include paint spraying, industrial and chemical processing, asbestos inspection, building cleaning, insulation laying, pesticide handling, powder coating and pharmaceutical manufacturing. Whilst also being super low linting, it is suitable for a wide range of applications including within many cleanrooms.
Colour: White
Certificeringer: Type 5/6, EN 1149-5, EN 1073-2, EN 14126; Cat. III.