Temperature probes, Lemo®, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P1720-16 P1760-20 P1760-25 P1760-15 P1760-30 P1720-24 P1760-10 P1720-32
BOHLP1720-16EA 1520 DKK
BOHLP1720-16 BOHLP1760-20 BOHLP1760-25 BOHLP1760-15 BOHLP1760-30 BOHLP1720-24 BOHLP1760-10 BOHLP1720-32
Temperature probes, Lemo®, BOLA
Temperatur prober
Used for temperature measurement in aggressive liquids and cable provides flexible connection from measuring device to medium.
  • Thermocouple in a PTFE-encapsulated stainless steel tube
  • Temperature probe of tip Ø 6mm and collar ring Ø 12mm
  • With white PTFE-coated cable (length of 1,5 m) and coupling (type Lemo® socket size 1, 4-pole)
  • Typical response times of T50 is 7 to 12 seconds and T90 is 14 to 16 seconds