Stirring shafts, moon shaped, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C376-02 C376-04 C376-06 C376-08 C376-10 C376-12 C376-14 C376-16 C376-18 C376-20 C376-22 C376-19 C376-64 C376-58
BOHLC376-02EA 1990 DKK
BOHLC376-02 BOHLC376-04 441-2929 BOHLC376-08 BOHLC376-10 BOHLC376-12 442-9205 442-9203 442-9204 BOHLC376-20 BOHLC376-22 BOHLC376-19 BOHLC376-64 BOHLC376-58
Stirring shafts, moon shaped, BOLA
Omrørere Omrøreaksler
Stirrer shafts made of PTFE-jacketed stainless steel and the half-moon shaped stirrer blade made of PTFE.
  • Ideal for stirring in round-bottom flasks with ground joint necks
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
  • Tangential flow with little turbulence