VWR®, Calibration Weight Sets, Class E2
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
611-8722EA 4220 DKK
611-8722 611-8723 611-8724 611-8725 611-8726 611-8727 611-8728 611-8729 611-8731 611-8732 611-8733 611-8734 611-8735 611-8736 611-8737 611-8742 611-8743 611-8744 611-8745 611-8746 611-8747 611-8748 611-8749 611-8751 611-8752 611-8753 611-8754 611-8755 611-8756 611-8757 611-8454 611-8456 611-8455 611-8457 611-8458 611-8459 611-8460 611-4938 611-4939 611-4944 611-4945
VWR®, Calibration Weight Sets, Class E2
Lodder Kalibreringslodder
Weights made from solid, anti-magnetic, polished stainless steel. Suitable for high resolution electronic balances.
  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate
Conform to OILM R111 recommendations.
Certificeringer: Conform to OILM R111 recommendations.
Bestillingsoplysninger: All weight sets listed in the 'set contents' table are available with or without COFRAC certificate -More Product Information