VWR®, Magnetic Stirring Bar Retriever
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
442-4549EA 207 DKK
442-4549 442-4550 442-4551 442-4552
VWR®, Magnetic Stirring Bar Retriever
Omrørere Omrøreudstyr
PTFE covered, rare earth magnetic core. For the removal of stirring magnets from vessels of all kinds.
  • Very good chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)