Radiation storage boxes, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F249600000 F249600001 F249600002 F249600003
BELAF249600000EA 19950 DKK
BELAF249600000 BELAF249600001 BELAF249600002 BELAF249600003
Radiation storage boxes, SP Bel-Art
Bokse Beta/gammastrålingsbokse
Storage boxes, lead lined polyethylene, suitable for storing vials of isotopes, contaminated instruments and tools, radioactive samples, solutions, or waste.
  • Safeguard personnel from exposure to radiation or lead
  • Handle opening is 89×29 mm
It has 0,76 mm thick lead shield encased in 6,4 mm thick high-density polyethylene which helps contain radiation without lead exposure. Each box has a cover of the same lead/polyethylene construction with a 32 mm external overlap designed to prevent radiation escape.