Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
463201 463200 446471 446473
738-0753EA 4170 DKK
738-0753 738-0752 738-0750 738-0751
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Mikroplader Immunologi mikroplader
PS, 96 wells per frame. For fluorescence/luminescence-based immunoassays and binding assays, these modules have been designed using the standard 96 MicroWell™ LockWell™ format. Opaque white modules have a reflective surface and offer low cross talk and minimal background. Opaque black modules with light absorbing surface offer minimum cross talk and back scattered light.
  • Each well locks in the frame of the same height to prevent jamming in machines
  • Easy to handle
  • Uniform washing and reading
  • Certified binding homogeneity for all four surfaces
  • MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains; PolySorp™ surface has a high affinity for hydrophobic molecules
Total volume per well: 350 µl