Transfection reagent, Insect GeneJuice®
Leverandør: Merck Millipore (Novagen)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
71259-4 71259-5 71259-3
71259-4EA 4890 DKK
71259-4 71259-5 71259-3
Transfection reagent, Insect GeneJuice®
Transfektionsreagenser
Insect GeneJuice® Transfection Reagent is formulated to maximise transfection efficiency of Sf9 insect cells. The reagent features extremely low toxicity to the cells and can be used for both transient and stable transfections in serum-containing or serum-free media, and for co-transfection of transfer plasmids with linearised virus DNA for the production of recombinant baculoviruses.
Leveringsinformation: Supplied as a 2 mg/ml suspension in 20 mM MES, 150 mM NaCl, pH 6.2 buffer. One ml is sufficient for 12 or 125 transfections in 10 ml suspension culture flasks or 35 mm plates, respectively.