Insulinsprøjter, Omnifix® 100 Solo og Duo
Leverandør: B. Braun
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9161376V 9161708V
720-2558EA 552 DKK
720-2558 BRAU9161708V
Insulinsprøjter, Omnifix® 100 Solo og Duo
Sprøjter Sprøjter, medicinske
PP, three-piece syringe for injection of U-100 insulin (50 or 100 IU/ml) for veterinary use. Smooth action, natural latex-free plunger stoppers with double sealing ring.
  • Permanent, easy-to-read graduations
  • Safe plunger backstop
  • Centric catheter filling
Emballering: 100 units per box.