Single use syringes, 2-piece, Injekt® Solo
Leverandør: B. Braun
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4606027V 4606051V 4606108V 4606205V 4606728V 4606701V 4606710V 4606736V
720-2518EA 85.3 DKK
720-2518 720-2519 720-2520 720-2521 720-2524 720-2522 720-2523 720-2525
Single use syringes, 2-piece, Injekt® Solo
Sprøjter Sprøjter, medicinske
Single use syringes for manual use, with green plunger. Easy aspiration up to the maximum volume. Barrel made from PP and plunger from PE.
  • Highly transparent barrel
  • Black, permanent graduation for ideal readability
  • Graduation extends beyond nominal volume
  • Minimal residual volume
  • Safe plunger backstop
Emballering: Dispenser box of 100 units, individually wrapped in blisters.