Biopsy processing/embedding cassettes, Slimsette®, M510
Leverandør: Simport Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M510-9 M510-8 M510-10 M510-11 M510-12 M510-2 M510-3 M510-4 M510-5 M510-6 M510-7
720-1612EA 1440 DKK
720-1612 720-1611 720-1599 720-1600 720-1601 720-1605 720-1606 720-1607 720-1608 720-1609 720-1610
Biopsy processing/embedding cassettes, Slimsette®, M510
Histologikassetter Indstøbningskassetter, Biopsiprøver
Acetal polymer. Cassette design is similar to that of the M509 model but specially designed to hold biopsy specimens.
  • Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
  • Disposable plastic cassettes are highly resistant to histological solutions
  • Small, square openings for good fluid exchange
  • Integral, recessed lid
  • Front face inclined 45° to enable use with certain automatic labelling systems
W×D×H: 41×28,5×6 mm
Emballering: Each case contains three boxes holding 500 cassettes each.