Anaerobic systems, anaerobic jars
Leverandør: DON WHITLEY SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A00002 A00005
710-1213EA 12880 DKK
710-1213 710-0841
Anaerobic systems, anaerobic jars
Anaerobe systemer
These anaerobic jars are robustly constructed in stainless steel and are simple to use.
  • Patented valve system
  • Fitted with the Oxoid low temperature catalyst sachet
They can be used with either gas generating envelopes or cylinder gas and will vent automatically if excess pressure is applied.
Bestillingsoplysninger: A large range of accessories are available. Please contact your local Avantor office for further information.
Leveringsinformation: Supplied ready-to-use and complete with a comprehensive spares kit.