pH buffer solutions, Pure Water™ and pHISA™ Adjustor, Orion™
Leverandør: Thermo Orion
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
700702 700003 700902 700402
662-0976EA 4310 DKK
662-0976 ORIO700003 662-1142 662-0977
pH buffer solutions, Pure Water™ and pHISA™ Adjustor, Orion™
Buffere pH buffere
Buffers for calibration giving fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water, and process water.
  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample
  • For best results, a high quality pH electrode is recommended
pHISA™Adjustor additive is designed to increase sample ionic strength without changing the pH of the sample.