Buffer solutions, Certipur®, ready to use, 1 litre bottles, Supelco®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.09406.1000
1.09407.1000
1.09408.1000
1.09409.1000
1.09406.1000EA
356.78
DKK
1.09406.1000
1.09407.1000
1.09408.1000
1.09409.1000
Buffer solutions, Certipur®, ready to use, 1 litre bottles, Supelco®
Buffere
pH buffere
Materialer til præcis kalibrering og kontrol af pH-instrumenter.
- Traceable to NIST primary standard and PTB
- Uncertainty ±0,015 (except pH 10 ±0,05)
- Reference temperature: +25 °C
Emballering: 1 litre plastic bottle.