Slide staining system, StainTray™ M920
Leverandør: Simport Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M920-1 M921-1 M920-2 M921-2
631-1922EA 5290 DKK
631-1922 631-1924 631-1923 631-1925
Slide staining system, StainTray™ M920
Farvekar
StainTray M920 has a rigid base that is resistant to many chemicals (except chlorinated hydrocarbons). It can hold up to 20 slides on four plastic rails covered with a polymer strip for holding slides perfectly in place even if the tray is held at an angle.
  • Choice of two possible covers: Clear for visual examination/black for fluorescent work
  • Drain plug can be removed and emptied easily
  • Four rubber feet to increase base stability
  • Units are stackable to save space
  • Single-handed operation makes handling safe and easy
Suitable for routine stainingMore Product Information