Transport system for cooled samples, minivaqbox
Leverandør: DELTA T
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
V020010 V020011 V020012
478-9925EA 9440 DKK
478-9925 478-9926 478-9927
Transport system for cooled samples, minivaqbox
Bokse Kølere/Frysebokse/Poser
This transport system uses vacuum insulation technology and is ideal for lengthy refrigerated transport of specimens, cells, and other sensitive items. The box is provided with a shoulder strap to make carrying easier. Use of the MiniVaqBox transport system with Delta T elements will ensure proper temperature maintenance without the application of any external energy for a period of 48 hours. First, the elements must be pre-cooled for a certain period of time until the fluid is solid. This will occur when the temperature is approximately 5 °C below the fluid’s melting point. The pre-cooling time depends on the number of fluid elements present.
  • The internal polyethylene layer in the box is easy to clean
Total temperature control for 48 h and more is made possible by the vacuum isolation panels and the phase change properties of the TempShell elements. The TempShell elements fit in the box exactly to ensure optimum insulation. The box can hold 5 TempShells or 1 TempShell with up to 8 TempFrames.
Interior volume: 16 litres
Weight 4,5 kg
Exterior W×D×H: 410×240×350 mm
Interior W×D×H: 300×200×330 mm