Tilbehør til digitale bloktermostater, QBA, QBD, QBH
Leverandør: GRANT INSTRUMENTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
QBL4 QBL2 QBL1 QBEP
460-0096EA 1490 DKK
460-0096 460-0095 460-0094 460-0093
Tilbehør til digitale bloktermostater, QBA, QBD, QBH
Bloktermostater Digitale bloktermostater
Tough clear polycarbonate covers maintain visibility while preventing anyone from accidentally touching a hot block.
  • Protect against splashing and are particularly recommended for use during operation at high temperatures