Measuring beaker with handle and lid
Leverandør: VITLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
97891 978941 97791 978081
216-3002EA 179 DKK
216-3002 VITL978941 VITL97791 VITL978081
Measuring beaker with handle and lid
Bægre
Collection vessel, made of SAN, clear or PP, translucent, for a wide variety of uses.
  • Embossed scale
  • With stable, easy-grip handle
  • White PC lid