Biological detergent concentrate, rea-clean®
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9790100 9790101 9790105 9790130
148-4230EA 139 DKK
148-4230 148-4231 148-3232 HIRS9790130
Biological detergent concentrate, rea-clean®
Renseopløsninger Detergenter og Overflade Rengøringsmidler
For all metal surfaces and plastic materials either in the concentrated or diluted solution.
  • Strong degreaser
  • Phosphate-free