VWR®, Block Weights, Iron, Class M2
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
611-8383EA 739 DKK
611-8383 611-8384 611-8385 611-8386 611-8387 611-8388 611-8389 611-8390
VWR®, Block Weights, Iron, Class M2
Lodder Kalibreringslodder
Rectangular weights made from cast iron, permanently magnetised.
  • Indelible unique identification number guarantees traceability
  • Painted grey
  • Adjustment cavity sealed with a lead plug
Certificeringer: Conforms to OIML, COFRAC accredited.