VWR®, Calibration Weights, Class M1
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
611-8253EA 102 DKK
611-8253 611-8254 611-8255 611-8256 611-8257 611-8258 611-8259 611-8260 611-8261 611-8262 611-8263 611-8264 611-8265 611-8266 611-8267 611-8268 611-8269 611-8270 611-8271 611-8272 611-8273 611-8274 611-8275 611-8276 611-8277 611-8278 611-8279 611-8280 611-8281 611-8282 611-8283 611-8284 611-8285 611-8286 611-8287 611-8288 611-8289 611-8294 611-8295 611-8296 611-8297 611-8228 611-8229 611-8230
VWR®, Calibration Weights, Class M1
Lodder Kalibreringslodder
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Leveringsinformation: All weights are supplied in plastic cases.