Capillary electrophoresis tubing
Leverandør: Trajan Scientific and Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
062801 062803 062812 062813 062823 062881
SGEA062801EA 421 DKK
SGEA062801 SGEA062803 SGEA062812 SGEA062813 SGEA062823 SGEA062881
Capillary electrophoresis tubing
Slanger Kromatografirør
Manufactured from precision bore tubing.
  • Hydrophobic and hydrophilic surface treatments
  • Wide pH range
  • 30, 50, 75 and 100 µm Int.Ø
  • Column-to-column reproducibility