SP Bel-Art® long-handled dippers
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F367810032 F367810016 F367800016 F367800032 F367820016 F367820032
BELAF367810032EA 2100 DKK
BELAF367810032 BELAF367810016 BELAF367800016 BELAF367800032 BELAF367820016 BELAF367820032
SP Bel-Art® long-handled dippers
Prøvetagere Prøveøser
These high density PP dippers will not rust or corrode.
  • Chemical resistance
  • Wooden dowel inside the handles for strength and rigidity
  • Welded handle to the rigid bowl at a 45° angle
Suitable for easy sampling in tanks, vats, lakes.
Bestillingsoplysninger: Handle (370 cm) is available either in one piece or two pieces screw-together construction.