Screw caps for scintillation vials, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
240917 241017 241009 240804 240805 240817 241018 241317 240818
548-2037EA 6260 DKK
548-2037 WHEA241017 WHEA241009 WHEA240804 WHEA240805 WHEA240817 WHEA241018 WHEA241317 WHEA240818
Screw caps for scintillation vials, WHEATON®
Låg Flaskelåg
White polypropylene construction.
  • Good chemical resistance
  • Good temperature resistance
Screw caps are designed for use with Wheaton glass or plastic liquid scintillation vials.