Magnetic stirring bars, PTFE, large cylindrical, Spinbar®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F371180006 F371180004
BELAF371180006EA 3290 DKK
BELAF371180006 BELAF371180004
Magnetic stirring bars, PTFE, large cylindrical, Spinbar®, SP Bel-Art
Omrørere Omrøremagneter
Spinbar® magnetic stirring bars with Alnico V magnet encapsulated in PTFE. The multifaceted stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
  • Chemical resistant
  • Will fit into vessels with a minimum neck diameter of 32 mm
  • Without pivot ring
  • Autoclave safe
Ideal for stirring large volumes of fluid in vessels on large magnetic stirrers or other large equipment.
Certificeringer: Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification