Dropping bottles, LDPE, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F116370001 F116370016 F116370002 F116370006 F116370008
BELAF116370001EA 495 DKK
BELAF116370001 BELAF116370016 BELAF116370002 BELAF116370006 BELAF116370008
Dropping bottles, LDPE, SP Bel-Art
Flasker Dråbeflasker
LDPE, translucent.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Flexible
The bottles and spouts have sealer caps that snap on easily for a tight seal. Spout openings are standard 0,76 mm. Opening can be enlarged by cutting back with scissors.