VWR® Traceable®, Waterproof Thermometers
Leverandør: VWR Collection
Ratings:
Gennemsntilig vurdering: 5.0 (Bedømmelse)
Samlet vurdering
Gennemsntilig vurdering
Var denne bedømmelse brugbar? Ja Nej
Ja: 3 Nej: 2 
Pharma / Techniker / Frankfurt
Ja, jeg anbefaler
Tolles Produkt, super Service, gerne wieder! Viele Grüße, VWR.com
Svar
vwr.com medarbejder Offentliggjort 01/04/2025
Danke für Ihr Feedback.
620-2092EA 460 DKK
620-2092 620-2099
VWR® Traceable®, Waterproof Thermometers
Termometre Digital termometre
This waterproof and shockproof thermometer is a safe replacement for any hazardous mercury instrument. The probe is resistant to most chemicals.
  • Waterproof construction for wet labs, wash-down areas and all field applications
  • Memory recalls maximum/minimum temperatures
  • Reading updates every second
Stainless steel probe (Lר: 203×3,5 mm).
Leveringsinformation: Supplied with probe, probe cover, silver oxide battery and Traceable® certificate.