VWR® Traceable®, Digital Dial Thermometers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-2082EA 664 DKK
620-2082 620-2095 620-2024
VWR® Traceable®, Digital Dial Thermometers
Termometre Digital termometre
These thermometers are safer than mercury thermometers. The smooth probe (620-2024) is suitable for viscous solutions or semi-solids, thanks to its piercing tip.
  • Readings updated every second
  • Large, bright LCD for easy reading
  • 620-2082 with metal disc (Ø: 12,7 mm) for surface measurements
Stainless steel probe (Lר: 133×3,8 mm).
Leveringsinformation: Supplied with silver-oxide battery and Traceable certificate of calibration.