2,2'-Bithiophen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
2,2′-Dithienyl, 2,2′-Bithienyl
B1276-1G
B1276-25G
TCIAB1276-1GEA
132
DKK
TCIAB1276-1G
TCIAB1276-25G
2,2'-Bithiophen ≥98.0% (by GC)
2,2'-Bithiophen
Formel:
C₈H₆S₂ Kogepunkt: 260°C |
MDL Number:
MFCD00005414 CAS-nummer: 492-97-7 EINECS: 207-767-2 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow to Green powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...