2,5-Dimercapto-1,3,4-thiadiazol dikaliumsalt ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L11105.06
L11105.14
L11105.06EA
432
DKK
L11105.06
L11105.14
2,5-Dimercapto-1,3,4-thiadiazol dikaliumsalt ≥97%
2,5-Dimercapto-1,3,4-thiadiazol dikaliumsalt
Purity: 97%
CAS No.: 4628-94-8
Molecular Formula: C2K2N2S3
Formula Weight: 226.43
Melting Point: ca 274° dec.
Beilstein Reference: 4917786
Formel:
C₂K₂N₂S₃ Smeltepunkt: ∼ 274 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003104 CAS-nummer: 4628-94-8 EINECS: 225-043-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...