N-Ethylrhodanin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Ethylrhodanine, 3-Ethyl-2-thioxothiazolidin-4-one

B23047.06 B23047.14
B23047.06EA 864 DKK
B23047.06 B23047.14
N-Ethylrhodanin ≥98%
N-Ethylrhodanin

Purity: 98% CAS No.: 7648-01-3 Molecular Formula: C5H7NOS2 Formula Weight: 161.25 Flash Point: 110°(230°F) Melting Point: 36-39° Boiling Point: 127-129°/4mm MDL No.: MFCD00051339

Formel: C₅H₇NOS₂
Kogepunkt: 127…129 °C (4 mmHg)
Smeltepunkt: 36…39 °C
Massefylde: 1,303
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051339
CAS-nummer: 7648-01-3
EINECS: 231-603-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR