O-Phenyl chlorthioformat ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Phenyl chlorothionocarbonate, Phenyl chlorothionoformate, O-Phenyl carbonochloridothioate
Purity: 98+%
CAS No.: 1005-56-7
Molecular Formula: C7H5ClOS
Formula Weight: 172.63
Storage Temperature: 2°C to 8°C
Flash Point: 81°(177°F)
Refractive Index: 1.5804
Odour: Acrid
Boiling Point: 81-83°/6mm
Beilstein Reference: 774830
UN No.: 3265
MDL No.: MFCD00004920
Formel:
C₇H₅ClOS Kogepunkt: 81…83 °C (6 mmHg) Massefylde: 1,248 Flammepunkt: 81 °C (177 °F) |
MDL Number:
MFCD00004920 CAS-nummer: 1005-56-7 EINECS: 213-736-4 UN nummer: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...