2-Methyl-1,3-dioxolan ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98%
CAS No.: 497-26-7
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: -2°(28°F)
Refractive Index: 1.3980
Boiling Point: 80-83°
Beilstein Reference: 102520
UN No.: 1993
MDL No.: MFCD00003210
Formel:
C₄H₈O₂ Kogepunkt: 80…83 °C Massefylde: 0,982 Flammepunkt: –2 °C (28 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003210 CAS-nummer: 497-26-7 EINECS: 207-841-4 ADR: 3,II |