2-(2-Aminoethyl)-1,3-dioxolan ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A1176-1G
A1176-5G
TCIAA1176-1GEA
780
DKK
TCIAA1176-1G
TCIAA1176-5G
2-(2-Aminoethyl)-1,3-dioxolan ≥98.0% (by GC, titration analysis)
2-(2-Aminoethyl)-1,3-dioxolan
Formel:
C₅H₁₁NO₂ Kogepunkt: 75°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00142756 CAS-nummer: 5754-35-8 EINECS: 227-277-2 UN nummer: 2733 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...