Borstandard, 1g/L B i vand 1% ammoniumhydroxid (from B(OH₃)) Specpure® enkeltelement standard til ICP
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
13859.AE 13859.AD 13859.AP
13859.AEEA 946 DKK
13859.AE 13859.AD 13859.AP
Borstandard, 1g/L B i vand 1% ammoniumhydroxid (from B(OH₃)) Specpure® enkeltelement standard til ICP
Standarder Kationiske standarder Boron Standards
Molecular Formula: H₃BO₃ in 1% NH₄OH
Physical Form: Liquid
MDL No.: MFCD00134034
Certificeringer: Traceable to NIST