Borstandard, 10g/L B i vand 1% ammoniumhydroxid (from B(OH₃)) Specpure® enkeltelement standard til ICP
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
14370.AD
14370.AE
14370.AP
14370.ADEA
1100
DKK
14370.AD
14370.AE
14370.AP
Borstandard, 10g/L B i vand 1% ammoniumhydroxid (from B(OH₃)) Specpure® enkeltelement standard til ICP
Standarder
Kationiske standarder
Boron Standards
Molecular Formula: H₃BO₃ in 1% NH₄OH
Physical Form: Liquid
MDL No.: MFCD00134034
Certificeringer: Traceable to NIST