1-Amino-3,3-diethoxypropan 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
269850250
269851000
ACRO269850250EA
991
DKK
ACRO269850250
ACRO269851000
1-Amino-3,3-diethoxypropan 97%
1-Amino-3,3-diethoxypropane
Formel:
C₇H₁₇NO₂ Molekylvægt: 147.22 g/mol Kogepunkt: 72 °C (16hPa) Massefylde: 0.91 g/cm³ Flammepunkt: 78 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00142581 CAS-nummer: 41365-75-7 UN nummer: 2735 ADR: 8 I |
Specifikationer Testresultater
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4233 to 1.4253 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...