trans-9-Octadecensyre

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (E)-Octadec-9-enoic acid

90250-100 90250-250 90250-50 90250-1
CAYM90250-100EA 536 DKK
CAYM90250-100 CAYM90250-250 CAYM90250-50 CAYM90250-1
trans-9-Octadecensyre
trans-9-Octadecensyre
Formel: C₁₈H₃₄O₂
MDL Number: MFCD00063954
CAS-nummer: 112-79-8
EINECS: 204-006-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR